Learn More
Medchemexpress LLC MOPS | 1132-61-2 | 100.0% | 1 ML
SDP
Catalog No. 5000410407
Encompass_Preferred
MOPS is a buffering agent utilized in biological applications. It effectively maintains the pH of mammalian cell culture media and significantly enhances the thermal stability of BSA (Bovine Serum Albumin).
  • Buffering agent for biological applications.
  • Maintains pH of mammalian cell culture media.
  • Enhances thermal stability of BSA.
Catalog No. 50-004-10407 Supplier Medchemexpress LLC Supplier No. HYD085910MM/1ML
MOPS is a buffering agent utilized in biological applications. It effectively maintains the pH of mammalian cell culture media and significantly enhances the thermal stability of BSA (Bovine Serum Albumin).
  • Buffering agent for biological applications.
  • Maintains pH of mammalian cell culture media.
  • Enhances thermal stability of BSA.