Learn More
Medchemexpress LLC MOPS | 1132-61-2 | 99.95% | 10 G
SDP
Catalog No. 5000410174
Encompass_Preferred
MOPS is a buffering agent used in biological applications. It functions to maintain the pH of mammalian cell culture media. Additionally, it has been shown to significantly enhance the thermal stability of BSA.
  • Functions as a buffering agent in biology.
  • Maintains the pH of mammalian cell culture media.
  • Significantly improves the thermal stability of BSA.
Catalog No. 50-004-10174 Supplier Medchemexpress LLC Supplier No. HYD085910G
MOPS is a buffering agent used in biological applications. It functions to maintain the pH of mammalian cell culture media. Additionally, it has been shown to significantly enhance the thermal stability of BSA.
  • Functions as a buffering agent in biology.
  • Maintains the pH of mammalian cell culture media.
  • Significantly improves the thermal stability of BSA.