Learn More
Alkali Scientific MOPS [3 (N morpholino) propanesulfonic acid], 5 Kg
SDP
Catalog No. 5000661639
Encompass_Preferred
MOPS (3-(N-morpholino)propane-1-sulfonic acid) is a pH buffering biochemical reagent commonly used in laboratory applications. This 5kg package of MOPS is ideal for preparing large volumes of buffer solutions, especially for high-throughput research and industrial applications. MOPS is widely used in molecular biology, protein chemistry, and enzyme studies, particularly in the pH range of 6.5 to 7.9. The large packaging size is suitable for laboratories that require consistent pH control in large-scale processes such as protein purification, electrophoresis, and DNA/RNA analysis. MOPS is essential for stabilizing biological macromolecules in research and industrial labs.
Catalog No. 50-006-61639 Supplier Alkali Scientific Supplier No. MOP008CP
MOPS (3-(N-morpholino)propane-1-sulfonic acid) is a pH buffering biochemical reagent commonly used in laboratory applications. This 5kg package of MOPS is ideal for preparing large volumes of buffer solutions, especially for high-throughput research and industrial applications. MOPS is widely used in molecular biology, protein chemistry, and enzyme studies, particularly in the pH range of 6.5 to 7.9. The large packaging size is suitable for laboratories that require consistent pH control in large-scale processes such as protein purification, electrophoresis, and DNA/RNA analysis. MOPS is essential for stabilizing biological macromolecules in research and industrial labs.