Learn More
Alkali Scientific MOPS [3 (N morpholino) propanesulfonic acid], 50 G
SDP
Catalog No. 5000660032
Encompass_Preferred
MOPS (3-(N-morpholino)propane-1-sulfonic acid) is a biochemical reagent widely used in laboratory applications for pH buffering. This 50g package of MOPS is commonly used for preparing small to medium quantities of buffer solutions for molecular biology, protein chemistry, and electrophoresis. The reagent is effective in maintaining a stable pH range of 6.5 to 7.9, which is essential for stabilizing proteins, nucleic acids, and enzymes during biochemical assays. MOPS is an important component in buffer systems for electrophoresis, chromatography, and enzyme studies where consistent pH is necessary to ensure reliable experimental outcomes.
Catalog No. 50-006-60032 Supplier Alkali Scientific Supplier No. MOP006CP
MOPS (3-(N-morpholino)propane-1-sulfonic acid) is a biochemical reagent widely used in laboratory applications for pH buffering. This 50g package of MOPS is commonly used for preparing small to medium quantities of buffer solutions for molecular biology, protein chemistry, and electrophoresis. The reagent is effective in maintaining a stable pH range of 6.5 to 7.9, which is essential for stabilizing proteins, nucleic acids, and enzymes during biochemical assays. MOPS is an important component in buffer systems for electrophoresis, chromatography, and enzyme studies where consistent pH is necessary to ensure reliable experimental outcomes.