Learn More
Alkali Scientific MOPS [3 (N morpholino) propanesulfonic acid], 500 G
SDP
Catalog No. 5000660062
Encompass_Preferred
MOPS (3-(N-morpholino)propane-1-sulfonic acid) is a versatile biochemical reagent used to maintain pH stability in laboratory experiments. This 500g package is suitable for medium to high-volume buffer preparations required in molecular biology, protein analysis, and biochemical assays. MOPS is commonly used in electrophoresis, chromatography, and enzyme assays where maintaining a consistent pH in the range of 6.5 to 7.9 is essential. It is particularly useful in applications that involve the stabilization of proteins, enzymes, and nucleic acids, making it a critical reagent in molecular research, cell biology, and biochemistry laboratories.
Catalog No. 50-006-60062 Supplier Alkali Scientific Supplier No. MOP007CP
Add to Cart
Add to Cart
MOPS (3-(N-morpholino)propane-1-sulfonic acid) is a versatile biochemical reagent used to maintain pH stability in laboratory experiments. This 500g package is suitable for medium to high-volume buffer preparations required in molecular biology, protein analysis, and biochemical assays. MOPS is commonly used in electrophoresis, chromatography, and enzyme assays where maintaining a consistent pH in the range of 6.5 to 7.9 is essential. It is particularly useful in applications that involve the stabilization of proteins, enzymes, and nucleic acids, making it a critical reagent in molecular research, cell biology, and biochemistry laboratories.