Learn More
Sigma Aldrich Fine Chemicals Biosciences MOPS 99.5 titration25KG
Catalog No. 502907430
Encompass
3-(N-Morpholino)propane sulfonic acid (MOPS) is an N-substituted amino sulfonic acid with a morpholinic ring. MOPS is capable of buffering within a pH range of 6.5-7.9. MOPS is widely used in biological and biochemical studies due to its inert properties. It does not interact with any metal ions in solutions and has significant metal-buffer stability especially with copper (Cu) nickel (Ni) manganese (Mn) zinc (Zn) cobalt (Co) ions. MOPS buffer maintains the pH of the mammalian cell culture medium. MOPS functions to maintain pH in denaturing gel electrophoresis of RNA. MOPS can modify lipid interactions and influence the thickness and barrier properties of membranes. MOPS interacts with bovine serum albumin and stabilizes the protein. Hydrogen peroxide oxidizes MOPS slowly to N-oxide form.
Catalog No. 50-290-7430 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M125425KG
Add to Cart
Add to Cart
Request a Quote
3-(N-Morpholino)propane sulfonic acid (MOPS) is an N-substituted amino sulfonic acid with a morpholinic ring. MOPS is capable of buffering within a pH range of 6.5-7.9. MOPS is widely used in biological and biochemical studies due to its inert properties. It does not interact with any metal ions in solutions and has significant metal-buffer stability especially with copper (Cu) nickel (Ni) manganese (Mn) zinc (Zn) cobalt (Co) ions. MOPS buffer maintains the pH of the mammalian cell culture medium. MOPS functions to maintain pH in denaturing gel electrophoresis of RNA. MOPS can modify lipid interactions and influence the thickness and barrier properties of membranes. MOPS interacts with bovine serum albumin and stabilizes the protein. Hydrogen peroxide oxidizes MOPS slowly to N-oxide form.