Learn More
Electron Microscopy Sciences Mops, Biological Buffer, 6.5-7.9 pH 100 GR
SDP
Catalog No. 5027420
Encompass_Preferred
Our buffers are ideal for maintaining a constant pH during fixation and have an ionic constitution to prevent the extraction of cellular components or the deposition of precipitates of fixative chemicals. The buffers are prepared fresh for each order and conveniently packaged. Desired pH should be specified when ordering.
pH: 6.5-7.9
Catalog No. 50-274-20 Supplier Electron Microscopy Sciences Supplier No. 18900
Our buffers are ideal for maintaining a constant pH during fixation and have an ionic constitution to prevent the extraction of cellular components or the deposition of precipitates of fixative chemicals. The buffers are prepared fresh for each order and conveniently packaged. Desired pH should be specified when ordering.
pH: 6.5-7.9