Learn More
Biotang Inc MOPS Buffered Saline 5X, pH 7.0. 125 mM MOPS, 750 mM Sodium Chloride, pH 7.0. Prepared in 18.2 megohms water and filtered through 0.22 micron filter. BB-570.
Catalog No. 50155810 Shop All BIOTANG Inc Products
Encompass_Preferred
MOPS Buffered Saline 5X, pH 7.0. 125 mM MOPS, 750 mM Sodium Chloride, pH 7.0. Prepared in 18.2 megohms water and filtered through 0.22 micron filter. BB-570.
Catalog No. 50-155-810 Supplier Biotang Inc Supplier No. BTBB461
MOPS Buffered Saline 5X, pH 7.0. 125 mM MOPS, 750 mM Sodium Chloride, pH 7.0. Prepared in 18.2 megohms water and filtered through 0.22 micron filter. BB-570.