Learn More
Kd Medical Inc MOPS BUFFER 10X, 1L
SDP
Catalog No. 501031430
Encompass_Preferred
10 x MOPS Buffer is intended for use at 1x as the electrophoresis and gel running buffer during the separation of RNA on denaturing formaldehyde/agarose gels. One 1 liter bottle.
Catalog No. 50-103-1430 Supplier Kd Medical Inc Supplier No. RGF6170
Add to Cart
Add to Cart
10 x MOPS Buffer is intended for use at 1x as the electrophoresis and gel running buffer during the separation of RNA on denaturing formaldehyde/agarose gels. One 1 liter bottle.