Learn More
Sigma Aldrich Fine Chemicals Biosciences MOPS SDS running buffer powder for mPAGE(R) Bis-Tris gels pack of 5 ea (Each packet makes 1 liter of running buffer) |
Catalog No. NC2186211
Encompass
MOPS SDS running buffer powder for mPAGE(R) Bis-Tris gels pack of 5 ea (Each packet makes 1 liter of running buffer) |
This item is not returnable. View return policy
Catalog No. NC2186211 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MPM0PS
MOPS SDS running buffer powder for mPAGE(R) Bis-Tris gels pack of 5 ea (Each packet makes 1 liter of running buffer) |