Learn More
Sigma Aldrich Fine Chemicals Biosciences MOPS-EDTA-Sodium Acetate M1L
Catalog No. 502904701
Encompass
MOPS - EDTA - Sodium Acetate (MESA) is the most commonly used running buffer for RNA electrophoresis prior to Northern blotting procedures. Applied voltage of 5 V/cm is recommended for maximum reSOLUTIONon.
Catalog No. 50-290-4701 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M57551L
MOPS - EDTA - Sodium Acetate (MESA) is the most commonly used running buffer for RNA electrophoresis prior to Northern blotting procedures. Applied voltage of 5 V/cm is recommended for maximum reSOLUTIONon.