Learn More
AG Scientific Inc MOPS Free Acid 50 G
Catalog No. 502268333
Encompass_Preferred
MOPS, also known as 3-(N-morpholino)propanesulfonic acid, is one of Good’s buffers. It is N-substituted amino sulfonic acid with a morpholinic ring and has zwitterionic and inert properties. MOPS buffer is broadly used in biological and biochemical studies. It can be mixed with MOPS sodium salt to attain desired pH.MOPS is recommended as an agent for non-coordinating buffers in solutions with metal ions because it hardly forms a complex with most metal ions such as Cu, Ni, Mn, Zn and Co ions. The Good's buffer interferes with the Folin protein assay and partially decomposes when autoclaved in the presence of glucose. MOPS can be used as a Good’s buffer because it has low UV absorptivity, minimal reactivity, stable pH and is soluble in water.
Catalog No. 50-226-8333 Supplier AG Scientific Inc Supplier No. M272150GM
MOPS, also known as 3-(N-morpholino)propanesulfonic acid, is one of Good’s buffers. It is N-substituted amino sulfonic acid with a morpholinic ring and has zwitterionic and inert properties. MOPS buffer is broadly used in biological and biochemical studies. It can be mixed with MOPS sodium salt to attain desired pH.MOPS is recommended as an agent for non-coordinating buffers in solutions with metal ions because it hardly forms a complex with most metal ions such as Cu, Ni, Mn, Zn and Co ions. The Good's buffer interferes with the Folin protein assay and partially decomposes when autoclaved in the presence of glucose. MOPS can be used as a Good’s buffer because it has low UV absorptivity, minimal reactivity, stable pH and is soluble in water.