Learn More
Gold Biotechnology Inc MOPS Free Acid Ultra Pure
SDP
Catalog No. 5000562044
Encompass_Preferred
MOPS is a buffering agent used in biochemistry and molecular biology that was selected and described by Good et al It is a zwitterionic morpholinic buffer that is useful for a pH range of 6 5 7 9 and commonly used for cell culture media as a running buffer in electrophoresis and for protein purification in chromatography MOPS lacks the ability to form a complex with most metal ions and is recommended for use as a non-coordinating buffer in solutions with metal ions
Catalog No. 50-005-62044 Supplier Gold Biotechnology Inc Supplier No. M790500
Add to Cart
Add to Cart
MOPS is a buffering agent used in biochemistry and molecular biology that was selected and described by Good et al It is a zwitterionic morpholinic buffer that is useful for a pH range of 6 5 7 9 and commonly used for cell culture media as a running buffer in electrophoresis and for protein purification in chromatography MOPS lacks the ability to form a complex with most metal ions and is recommended for use as a non-coordinating buffer in solutions with metal ions