Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOPS Hemisodium Salt, Biological Buffer, Spectrum™ Chemical
SDP

Catalog No. 18611818 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
10 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-818 100 g Poly Bottle
18-611-820 10 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-611-818 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M3003100GM
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 117961-20-3
Molecular Formula C14H29N2NaO8S2
Molecular Weight (g/mol) 440.50
InChI Key OVCWZKQPQOVIDT-UHFFFAOYSA-M
IUPAC Name sodium 3-(morpholin-4-yl)propane-1-sulfonic acid 3-(morpholin-4-yl)propane-1-sulfonate
SMILES [Na+].OS(=O)(=O)CCCN1CCOCC1.[O-]S(=O)(=O)CCCN1CCOCC1

Specifications

CAS Max % 1
pH 6.7 to 7.3
Physical Form Crystalline Powder
Quantity 100 g
Packaging Poly Bottle
Percent Purity 100%
Grade Reagent
Chemical Name or Material MOPS Hemisodium Salt

CAUTION: For use by qualified personnel for research and development or other authorized use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.