Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOPS Hemisodium Salt, Biological Buffer, Spectrum™ Chemical
SDP

Catalog No. 18611820 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 g
10 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-820 10 kg Poly Bottle
18-611-818 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-611-820 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M300310KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 117961-20-3
Molecular Formula C14H29N2NaO8S2
Molecular Weight (g/mol) 440.50
InChI Key OVCWZKQPQOVIDT-UHFFFAOYSA-M
IUPAC Name sodium 3-(morpholin-4-yl)propane-1-sulfonic acid 3-(morpholin-4-yl)propane-1-sulfonate
SMILES [Na+].OS(=O)(=O)CCCN1CCOCC1.[O-]S(=O)(=O)CCCN1CCOCC1

Specifications

CAS Max % 1
For Use With (Application) Molecular Biology
pH 6.7 to 7.3
Physical Form Crystalline Powder
Quantity 10 kg
Solution Type Buffer
Packaging Poly Bottle
Percent Purity 100%
Grade Reagent
Chemical Name or Material MOPS Hemisodium Salt

CAUTION: For use by qualified personnel for research and development or other authorized use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.