Learn More
Genscript Corporation Tris-MOPS-SDS Running Buffer Powder
Catalog No. NC1278898
Encompass
Tris-MOPS-SDS Running Buffer Powder are used for ExpressPLUS and SurePAGE gel transfer. Using proprietary techniques, Tris-MOPS-SDS Running Buffer Powder are made to have long shelf life, high resolution, fast electrophoresis, smaller volume.
Catalog No. NC1278898 Supplier Genscript Corporation Supplier No. M00138
Tris-MOPS-SDS Running Buffer Powder are used for ExpressPLUS and SurePAGE gel transfer. Using proprietary techniques, Tris-MOPS-SDS Running Buffer Powder are made to have long shelf life, high resolution, fast electrophoresis, smaller volume.