Learn More
National Diagnostics MOPS-SDS Running Buffer (20X), 1L
SDP
Catalog No. NC1226207
Encompass
Reliable and Economical, 18 Megohm Water, and 0.2 Micron Filtered. Ideal running buffer for high resolution SDS PAGE of proteins. 20X concentrate makes a working solution of 50mM Tris MOPS, 0.1% SDS and 1mM EDTA.
Catalog No. NC1226207 Supplier National Diagnostics Supplier No. EC8671L
Reliable and Economical, 18 Megohm Water, and 0.2 Micron Filtered. Ideal running buffer for high resolution SDS PAGE of proteins. 20X concentrate makes a working solution of 50mM Tris MOPS, 0.1% SDS and 1mM EDTA.