Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals MOPS-SDS running buffer (20X)

Catalog No. AAJ62847K2
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62847K2 1 L
AAJ62847AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62847K2 Supplier Thermo Scientific Chemicals Supplier No. J62847K2
Add to Cart

MOPS-SDS running buffer (20X) is a buffer frequently used for separating medium- to large-sized proteins on a gel.

MOPS SDS Running Buffer is recommended for separating medium- to large-sized proteins. MOPS-SDS Running Buffer is suitable for protein gel electrophoresis. Use of MOPS buffer allows proteins to run slower than when using MES buffer. This buffer is used in 1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis - See more at:

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • MOPS-SDS running buffer is a zwitterionic, morpholinic buffer that allows the separation of medium- to large-sized proteins

Application

  • MOPS is used as a running buffer in electrophoresis. The use of MOPS buffer allows proteins to run slower than when MES buffer is used
  • MOPS-SDS running buffer, 20X concentrated, is formulated to be used for running proteins on an SDS gel
  • This buffer contains SDS detergent to maintain the denaturation of proteins and MOPS to buffer the pH
  • It is commonly used for cell culture media and protein purification in chromatography
  • MOPS is frequently applied in buffered culture media for bacteria, yeast, and mammalian cells
  • It is also applied in the bicinchoninic acid (BCA) assay
  • It is used in separating RNA in agarose gels
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material MOPS-SDS running buffer
Concentration 20X
Physical Form Liquid
Quantity 1 L
Solubility Information It is soluble in water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.