Learn More
Bioworld MOPS-SDS Running Buffer (20X), 500 mL
Catalog No. 50196661
Encompass_Preferred
MOPS-SDS Running Buffer for Protein Gel electrophoresisSDS running buffer with MOPS and Tris (pH 7.7)20X concentrationAll bioWORLD running buffers are prepared with high quality reagentsSpecifications:Ready to use, just dilute in distilled water. Save time and ensure reproducible results.1X contains: 50mM MOPS, 50mM Tris base, 1mM EDTA, and 0.1% SDS
Catalog No. 50-196-661 Supplier Bioworld Supplier No. 105300071
Add to Cart
Add to Cart
MOPS-SDS Running Buffer for Protein Gel electrophoresisSDS running buffer with MOPS and Tris (pH 7.7)20X concentrationAll bioWORLD running buffers are prepared with high quality reagentsSpecifications:Ready to use, just dilute in distilled water. Save time and ensure reproducible results.1X contains: 50mM MOPS, 50mM Tris base, 1mM EDTA, and 0.1% SDS