Learn More
Alkali Scientific MOPS, Sodium Salt [3 (N Morpholino) propanesulfonic acid sodium salt], 50 G
SDP
Catalog No. 5000660066
Encompass_Preferred
MOPS (3-(N-Morpholino)propanesulfonic acid sodium salt) is a versatile biochemical reagent used for pH buffering in various research applications. This 50g bottle provides a compact quantity of MOPS sodium salt, ideal for laboratories with moderate buffer preparation needs. MOPS is widely used in molecular biology, biochemistry, and biotechnology for stabilizing pH in enzyme reactions, protein assays, and cell culture. Its pH buffering capacity at a range of 7.0–7.5 is particularly beneficial in experiments where a consistent environment is essential. MOPS also serves as an essential reagent in electrophoresis, chromatography, and other scientific procedures that demand precise pH control.
Catalog No. 50-006-60066 Supplier Alkali Scientific Supplier No. MOP012CP
MOPS (3-(N-Morpholino)propanesulfonic acid sodium salt) is a versatile biochemical reagent used for pH buffering in various research applications. This 50g bottle provides a compact quantity of MOPS sodium salt, ideal for laboratories with moderate buffer preparation needs. MOPS is widely used in molecular biology, biochemistry, and biotechnology for stabilizing pH in enzyme reactions, protein assays, and cell culture. Its pH buffering capacity at a range of 7.0–7.5 is particularly beneficial in experiments where a consistent environment is essential. MOPS also serves as an essential reagent in electrophoresis, chromatography, and other scientific procedures that demand precise pH control.