Learn More
Research Products International Corp MOPS, Sodium Salt [3-(N-Morpholino)-propanesulfonic acid sodium salt], 500 Grams
SDP
Catalog No. 50488831
Encompass_Preferred
MOPS Sodium Salt, also known as 3-(N-Morpholino) propanesulfonic acid sodium salt, is a chemical compound that plays a significant role in the life science marketplace. It is a zwitterionic buffer commonly used in biological and biochemical research and is primarily utilized as a buffering agent to maintain a stable pH in various experimental procedures.
It is particularly effective in the pH range of 6.5 to 7.9, making it suitable for applications requiring slightly acidic to neutral pH conditions. MOPS Sodium Salt is commonly used in protein purification, enzyme assays, nucleic acid research, and electrophoresis techniques where a specific pH range needs to be maintained. Its buffering capacity and stability within the desired pH range make it valuable in these applications. 
Catalog No. 50-488-831 Supplier Research Products International Corp Supplier No. M92040500.0
MOPS Sodium Salt, also known as 3-(N-Morpholino) propanesulfonic acid sodium salt, is a chemical compound that plays a significant role in the life science marketplace. It is a zwitterionic buffer commonly used in biological and biochemical research and is primarily utilized as a buffering agent to maintain a stable pH in various experimental procedures.
It is particularly effective in the pH range of 6.5 to 7.9, making it suitable for applications requiring slightly acidic to neutral pH conditions. MOPS Sodium Salt is commonly used in protein purification, enzyme assays, nucleic acid research, and electrophoresis techniques where a specific pH range needs to be maintained. Its buffering capacity and stability within the desired pH range make it valuable in these applications. 