Learn More
Medchemexpress LLC MOPS (sodium salt) | 71119-22-7 | 99.8% | 1 KG
SDP
Catalog No. 5000410169
Encompass_Preferred
MOPS sodium salt is a buffering agent used in biology. It maintains the pH of mammalian cell culture media and significantly improves the thermal stability of BSA.
  • Buffering agent for biological applications.
  • Maintains pH in mammalian cell culture media.
  • Improves thermal stability of BSA.
Catalog No. 50-004-10169 Supplier Medchemexpress LLC Supplier No. HYD0859A1KG
MOPS sodium salt is a buffering agent used in biology. It maintains the pH of mammalian cell culture media and significantly improves the thermal stability of BSA.
  • Buffering agent for biological applications.
  • Maintains pH in mammalian cell culture media.
  • Improves thermal stability of BSA.