Learn More
Medchemexpress LLC MOPS (sodium salt) | 71119-22-7 | 99.8% | 25 G
SDP
Catalog No. 5000414466
Encompass_Preferred
MOPS sodium salt is a buffering agent utilized in biological applications. MOPS buffer is effective in maintaining the pH of mammalian cell culture media and has been shown to significantly improve the thermal stability of BSA. It is relatively safe for short-term experiments, though it may reduce cell viability after 4 hours, suggesting experimental duration limits of 4-12 hours.
  • Buffering agent for biological research.
  • Maintains pH in mammalian cell culture media.
  • Significantly improves the thermal stability of BSA.
  • Relatively safe for short-term experiments, though it may reduce cell viability after 4 hours, suggesting experimental duration limits of 4-12 hours.
  • Can be used in the preparation of keratinocyte and microbial culture media.
Catalog No. 50-004-14466 Supplier Medchemexpress LLC Supplier No. 25GHYD0859A
Add to Cart
Add to Cart
MOPS sodium salt is a buffering agent utilized in biological applications. MOPS buffer is effective in maintaining the pH of mammalian cell culture media and has been shown to significantly improve the thermal stability of BSA. It is relatively safe for short-term experiments, though it may reduce cell viability after 4 hours, suggesting experimental duration limits of 4-12 hours.
  • Buffering agent for biological research.
  • Maintains pH in mammalian cell culture media.
  • Significantly improves the thermal stability of BSA.
  • Relatively safe for short-term experiments, though it may reduce cell viability after 4 hours, suggesting experimental duration limits of 4-12 hours.
  • Can be used in the preparation of keratinocyte and microbial culture media.