Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MOPS sodium salt | 71119-22-7 | 99.8% | 500 G
SDP

Catalog No. 5000410602
Encompass_Preferred

MOPS sodium salt is a buffering agent used in biology. MOPS buffer maintains the pH of mammalian cell culture media. It significantly improves the thermal stability of BSA. MOPS sodium salt is relatively safe for short-term experiments, but it may significantly reduce cell viability after 4 hours, so it is recommended to limit the experiment duration to 4-12 hours.

  • Purity: 99.8%
  • Appearance: Solid
  • Color: White to off-white
  • Storage: 4°C, sealed, away from moisture. In solvent: -80°C for 6 months; -20°C for 1 month (sealed, away from moisture).
  • Solubility in H2O: 100 mg/mL (needs ultrasonic)
  • Solubility in DMSO: 50 mg/mL (needs ultrasonic; use newly opened DMSO)

Catalog No. 50-004-10602 Supplier Medchemexpress LLC Supplier No. HYD0859A500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MOPS sodium salt is a buffering agent used in biology. MOPS buffer maintains the pH of mammalian cell culture media. It significantly improves the thermal stability of BSA. MOPS sodium salt is relatively safe for short-term experiments, but it may significantly reduce cell viability after 4 hours, so it is recommended to limit the experiment duration to 4-12 hours.

  • Purity: 99.8%
  • Appearance: Solid
  • Color: White to off-white
  • Storage: 4°C, sealed, away from moisture. In solvent: -80°C for 6 months; -20°C for 1 month (sealed, away from moisture).
  • Solubility in H2O: 100 mg/mL (needs ultrasonic)
  • Solubility in DMSO: 50 mg/mL (needs ultrasonic; use newly opened DMSO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.