Learn More
Medchemexpress LLC Mops (sodium salt) | 71119-22-7 | 99.83% | 10 G
SDP
Catalog No. 5000409976
Encompass_Preferred
The product is a buffering agent utilized in biology. It serves to maintain the pH of mammalian cell culture media and notably enhances the thermal stability of BSA.
  • Functions as a buffering agent in biological applications.
  • Maintains the pH level of mammalian cell culture media.
  • Significantly improves the thermal stability of BSA.
  • Can be used in the preparation of keratinocyte and microbial culture medium.
  • Relatively safe for short-term experiments, though prolonged use may reduce cell viability.
Catalog No. 50-004-09976 Supplier Medchemexpress LLC Supplier No. HYD0859A10G
The product is a buffering agent utilized in biology. It serves to maintain the pH of mammalian cell culture media and notably enhances the thermal stability of BSA.
  • Functions as a buffering agent in biological applications.
  • Maintains the pH level of mammalian cell culture media.
  • Significantly improves the thermal stability of BSA.
  • Can be used in the preparation of keratinocyte and microbial culture medium.
  • Relatively safe for short-term experiments, though prolonged use may reduce cell viability.