Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOPSO, 99.5%, MP Biomedicals™

Catalog No. ICN15245580 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15245580 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15245580 Supplier MP Biomedicals, Inc Supplier No. 0215245580
Add to Cart
Add to Cart

Chemical Identifiers

CAS 79803-73-9
Molecular Formula C7H14NNaO5S
Molecular Weight (g/mol) 247.241
InChI Key WSFQLUVWDKCYSW-UHFFFAOYSA-M
Synonym sodium 2-hydroxy-3-morpholinopropane-1-sulfonate, mopso sodium salt, 3-morpholino-2-hydroxypropanesulfonic acid sodium salt, 3-n-morpholinyl-2-hydroxypropanesulfonic acid sodium salt, sodium 2-hydroxy-3-morpholin-4-yl propane-1-sulfonate, mopso sodium salt titration, mopso, 0.2m buffer solution, ph 6.0, mopso, 0.2m buffer solution, ph 6.5, mopso, 0.2m buffer solution, ph 7.0, mopso, 0.2m buffer solution, ph 7.5
IUPAC Name sodium;2-hydroxy-3-morpholin-4-ylpropane-1-sulfonate

Specifications

Color White
Physical Form Powder
Quantity 100 g
Formula Weight 247.2
Chemical Name or Material MOPSO
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.