Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MOPSO sodium | 79803-73-9 | 97.0% | 247.24 | C7H14NNaO5S | 500 G
SDP

Catalog No. 5000209734
Encompass_Preferred

MOPSO sodium is the sodium salt of MOPSO, a zwitterionic buffering agent used in biochemical and molecular biology applications to prepare buffer solutions.

  • Used for preparation of buffer solutions.
  • Zwitterionic buffering agent; effective pH range typically near physiological pH.
  • CAS number 79803-73-9.
  • Chemical formula C7H14NNaO5S; molecular weight 247.24.
  • Purity approximately 97.0% and available in 500 g packages.

Catalog No. 50-002-09734 Supplier Medchemexpress LLC Supplier No. HYW040173500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MOPSO sodium is the sodium salt of MOPSO, a zwitterionic buffering agent used in biochemical and molecular biology applications to prepare buffer solutions.

  • Used for preparation of buffer solutions.
  • Zwitterionic buffering agent; effective pH range typically near physiological pH.
  • CAS number 79803-73-9.
  • Chemical formula C7H14NNaO5S; molecular weight 247.24.
  • Purity approximately 97.0% and available in 500 g packages.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.