Learn More
Alkali Scientific MOPSO Sodium Salt, 1 Kg
SDP
Catalog No. 5000661604
Encompass_Preferred
MOPSO Sodium Salt, 1 Kg, is an essential buffering agent used to maintain pH stability in a variety of laboratory applications, especially in protein and nucleic acid studies. The MOPSO buffer provides reliable pH control in the range of 6.5 to 7.9, making it an ideal choice for enzyme assays, protein extractions, and other biochemical processes. The 1 kg size is perfect for large-scale experiments or labs that require frequent use of this buffer for ongoing research. MOPSO is also widely used in pharmaceutical and biotechnology labs, where consistent pH levels are crucial for reproducible results in complex biological assays.
Catalog No. 50-006-61604 Supplier Alkali Scientific Supplier No. PSO003CP
Add to Cart
Add to Cart
MOPSO Sodium Salt, 1 Kg, is an essential buffering agent used to maintain pH stability in a variety of laboratory applications, especially in protein and nucleic acid studies. The MOPSO buffer provides reliable pH control in the range of 6.5 to 7.9, making it an ideal choice for enzyme assays, protein extractions, and other biochemical processes. The 1 kg size is perfect for large-scale experiments or labs that require frequent use of this buffer for ongoing research. MOPSO is also widely used in pharmaceutical and biotechnology labs, where consistent pH levels are crucial for reproducible results in complex biological assays.