Learn More
Alkali Scientific MOPSO Sodium Salt, 250 G
SDP
Catalog No. 5000661267
Encompass_Preferred
MOPSO Sodium Salt, 250 G, is a highly effective buffering reagent used in laboratory applications requiring stable pH control. MOPSO (Morpholinopropanesulfonic Acid Sodium Salt) is particularly useful for maintaining pH stability in experiments involving enzymes, proteins, and nucleic acids. This buffer offers a pH range of 6.5 to 7.9, making it ideal for biological assays, protein extraction, and other biochemical procedures. The 250 g size is suitable for labs that require frequent use of MOPSO buffer and offers a cost-effective solution for maintaining consistent conditions across a variety of experiments.
Catalog No. 50-006-61267 Supplier Alkali Scientific Supplier No. PSO001CP
MOPSO Sodium Salt, 250 G, is a highly effective buffering reagent used in laboratory applications requiring stable pH control. MOPSO (Morpholinopropanesulfonic Acid Sodium Salt) is particularly useful for maintaining pH stability in experiments involving enzymes, proteins, and nucleic acids. This buffer offers a pH range of 6.5 to 7.9, making it ideal for biological assays, protein extraction, and other biochemical procedures. The 250 g size is suitable for labs that require frequent use of MOPSO buffer and offers a cost-effective solution for maintaining consistent conditions across a variety of experiments.