Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOPSO Sodium Salt, MP Biomedicals™

Catalog No. 0215245525 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-152-45525 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-152-45525 Supplier MP Biomedicals, Inc Supplier No. 0215245525
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MOPSO Sodium Salt is a zwitterionic aminosulfonate buffer.

  • MOPSO sodium salt is a zwitterionic aminosulfonate buffer that is very similar in structure to MOPS (3-morpholinopropanesulfonic acid), differing by the presence of a hydroxyl group on C-2 of the propane moiety.
  • It has been utilized as a buffer component in the analysis of copper by two distinct methods:
    • (1) A flow injection micellar technique of the catalytic reaction of the reaction between 3-methyl-2-benzothiazolinone hydrazone with N-ethyl-N-(2-hydroxy-3-sulfopropyl)-3,5-dimethoxyaniline.
    • (2) An electrospray-ionization quadrapole time-of-flight mass spectroscopy technique to measure the formation of chelating complexes of MOPSO with copper.
  • It has also been utilized as a component of buffered charcoal yeast extract medium.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79803-73-9
Molecular Formula C7H14NNaO5S
Molecular Weight (g/mol) 247.24
InChI Key WSFQLUVWDKCYSW-UHFFFAOYNA-M
IUPAC Name sodium 2-hydroxy-3-(morpholin-4-yl)propane-1-sulfonate
SMILES [Na+].OC(CN1CCOCC1)CS([O-])(=O)=O

Specifications

Melting Point 270°C to 290°C
Color White
pH (1% aq soln)
Physical Form Crystalline Powder
Quantity 25 g
Solution Type Buffer
Chemical Name or Material MOPSO Sodium Salt
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.