Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Moracin M | 56317-21-6 | 99.0% | 1 ML
SDP

Catalog No. 5000231024
Encompass_Preferred

Moracin M is a phenolic endogenous metabolite supplied for research and analytical applications (CAS 56317-21-6). It has molecular formula C14H10O4 and molecular weight 242.23 g/mol. Manufacturer documentation provides DMSO solubility guidance and example volumes for preparing 1 mM, 5 mM, and 10 mM stock solutions.

  • Analytical standard for research and analytical applications.
  • Reported phosphodiesterase inhibitory activity.
  • Molecular formula C14H10O4.
  • Molecular weight 242.23 g/mol.
  • Manufacturer provides DMSO solubility guidance and example volumes to prepare 1 mM, 5 mM, and 10 mM stocks.
  • Supplied as a solid with solution preparation guidance.

Catalog No. 50-002-31024 Supplier Medchemexpress LLC Supplier No. HY12294210MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Moracin M is a phenolic endogenous metabolite supplied for research and analytical applications (CAS 56317-21-6). It has molecular formula C14H10O4 and molecular weight 242.23 g/mol. Manufacturer documentation provides DMSO solubility guidance and example volumes for preparing 1 mM, 5 mM, and 10 mM stock solutions.

  • Analytical standard for research and analytical applications.
  • Reported phosphodiesterase inhibitory activity.
  • Molecular formula C14H10O4.
  • Molecular weight 242.23 g/mol.
  • Manufacturer provides DMSO solubility guidance and example volumes to prepare 1 mM, 5 mM, and 10 mM stocks.
  • Supplied as a solid with solution preparation guidance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.