Learn More
Medchemexpress LLC Moracin P | 102841-46-3 | 5 MG
SDP
Catalog No. 5000417855
Encompass_Preferred
Moracin P is a 2-arylbenzofuran isolated from the Mori Cortex Radicis. It exhibits potent in vitro inhibitory activity against hypoxia-inducible factor (HIF-1) and reduces oxygen-glucose deprivation (OGD)-induced reactive oxygen species (ROS) production. Moracin P has neuroprotective and anti-inflammatory effects, and enhances cell viability against OGD-induced cell death in neuroblastoma SH-SY5Y cells.
  • Potent in vitro inhibitory activity against hypoxia-inducible factor (HIF-1)
  • Reduces oxygen-glucose deprivation (OGD)-induced reactive oxygen species (ROS) production
  • Possesses neuroprotective effects
  • Possesses anti-inflammatory effects
  • Enhances cell viability against oxygen-glucose deprivation (OGD)-induced cell death
Catalog No. 50-004-17855 Supplier Medchemexpress LLC Supplier No. HYN32435MG
Moracin P is a 2-arylbenzofuran isolated from the Mori Cortex Radicis. It exhibits potent in vitro inhibitory activity against hypoxia-inducible factor (HIF-1) and reduces oxygen-glucose deprivation (OGD)-induced reactive oxygen species (ROS) production. Moracin P has neuroprotective and anti-inflammatory effects, and enhances cell viability against OGD-induced cell death in neuroblastoma SH-SY5Y cells.
  • Potent in vitro inhibitory activity against hypoxia-inducible factor (HIF-1)
  • Reduces oxygen-glucose deprivation (OGD)-induced reactive oxygen species (ROS) production
  • Possesses neuroprotective effects
  • Possesses anti-inflammatory effects
  • Enhances cell viability against oxygen-glucose deprivation (OGD)-induced cell death