Learn More
Medchemexpress LLC Moracin p | 102841-46-3 | 99.6% | 326.34 | C19H18O5 | 10 MG
SDP
Catalog No. 5000289497
Encompass_Preferred
Moracin P is a 2-arylbenzofuran isolated from Mori Cortex Radicis. It inhibits hypoxia-inducible factor (HIF-1), reduces oxygen-glucose deprivation (OGD)-induced reactive oxygen species (ROS), and demonstrates neuroprotective and anti-inflammatory effects - suitable for in vitro and in vivo research applications.
  • Demonstrates HIF-1 inhibitory activity.
  • Reduces ROS production in OGD models.
  • Exhibits neuroprotective effects.
  • Shows anti-inflammatory activity.
  • High purity (≈99.6%) and solid form; highly soluble in DMSO (≥100 mg/mL).
  • Stability: store protected from light at 4°C; in solution store at -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-89497 Supplier Medchemexpress LLC Supplier No. HYN324310MG
Moracin P is a 2-arylbenzofuran isolated from Mori Cortex Radicis. It inhibits hypoxia-inducible factor (HIF-1), reduces oxygen-glucose deprivation (OGD)-induced reactive oxygen species (ROS), and demonstrates neuroprotective and anti-inflammatory effects - suitable for in vitro and in vivo research applications.
  • Demonstrates HIF-1 inhibitory activity.
  • Reduces ROS production in OGD models.
  • Exhibits neuroprotective effects.
  • Shows anti-inflammatory activity.
  • High purity (≈99.6%) and solid form; highly soluble in DMSO (≥100 mg/mL).
  • Stability: store protected from light at 4°C; in solution store at -80°C (6 months) or -20°C (1 month).