Learn More
Medchemexpress LLC Morantel tartrate | 26155-31-7 | MFCD00079449 | 99.01% | 25 MG
SDP
Catalog No. 5000414430
Encompass_Preferred
Morantel tartrate is an anthelmintic agent used to control subclinical gastrointestinal parasitism among cattle on pasture. It can be found in ruminal, abomasal, ileal fluids, and feces for over 98 days after administration.
  • Controls subclinical gastrointestinal parasitism in cattle.
  • Effective against mature forms of various nematodes in cattle.
  • Used in other ruminant species.
Catalog No. 50-004-14430 Supplier Medchemexpress LLC Supplier No. 25MGHYB1073
Morantel tartrate is an anthelmintic agent used to control subclinical gastrointestinal parasitism among cattle on pasture. It can be found in ruminal, abomasal, ileal fluids, and feces for over 98 days after administration.
  • Controls subclinical gastrointestinal parasitism in cattle.
  • Effective against mature forms of various nematodes in cattle.
  • Used in other ruminant species.