Learn More
Selleck Chemical LLC Morantel tartrate
SDP
Catalog No. 5000003333
Encompass_Preferred
Morantel (CP-12009-18 UK 296418) is a positive allosteric modulator of neuronal nicotinic acetylcholine receptors (nAChRs)
Catalog No. 50-000-03333 Supplier Selleck Chemical LLC Supplier No. S57475MG
Morantel (CP-12009-18 UK 296418) is a positive allosteric modulator of neuronal nicotinic acetylcholine receptors (nAChRs)