Learn More
ATCC Bacillus anthracis; Bacteriophage WIP-1
Supplier:  ATCC BAA1254B2
Encompass
Bacillus anthracis bacteriophage WIP-1 is a virus that is propagated in the bacterium Bacillus anthracis. This bacteriophage should be kept frozen until used.
Catalog No. 50-238-0748