Learn More
ATCC Moraxella; (Moraxella); osloensis
Catalog No. 502384777
Encompass
Type strain that produces restriction endonuclease MosI.
Catalog No. 50-238-4777 Supplier ATCC Supplier No. 19976
Type strain that produces restriction endonuclease MosI.