Learn More
Aviva Systems Biology Corp MORC3 Antibody - N-terminal region (ARP45856_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45856_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MORC3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MORC3. Alternate names: NXP2, ZCW5, ZCWCC3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59154