Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Mordalphos Pd G4

Catalog No. 900276250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-027-6250MG 250 mg
90-027-61G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 90-027-6250MG Supplier Sigma Aldrich Supplier No. 900276250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

A powerful ligand for classic cross-coupling reactions combined with the Buchwald Fourth Generation Palladacycle. Bench stable, soluble in most organic solvents.

Specifications

CAS 1810068-30-4
Melting Point 160.3°C
Molecular Formula C34H29F12N2O2P
Linear Formula C34H29F12N2O2P
Quantity 250 mg
Synonym 2'-(Bis(3,5-bis(trifluoromethyl)phenyl)phosphanyl)-3',6'-dimethoxy-N ,N ,N ,N -tetramethyl-[1,1'-biphenyl]-2,6-diamine; 2'-(Bis(3,5-bis(trifluoromethyl)phenyl)phosphanyl)-3',6'-dimethoxyN2,N2,N6,N6 -tetramethyl-[1,1'-biphenyl]-2,6-diamine; 2-{Bis[3,5-bis(trifluoromethyl)phenyl]phosphino}-3,6-dimethoxy -2',6'-bis(dimethylamino)-1,1'-biphenyl
Molecular Weight (g/mol) 756.56 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.