Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mordant Black 11 Triturate R, 1% (w/w) in Sodium Chloride, Ricca Chemical
SDP

Catalog No. 5215100 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
White Poly Wide Mouth Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
5215-100 100 g White Poly Wide Mouth Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5215-100 Supplier Ricca Chemical Company Supplier No. 5215100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mordant black triturate indicator reagent 1% w/w in sodium chlorideis suitabtle for laboratory applications.

Chemical Identifiers

CAS 7647-14-5 , 1787-61-7
Molecular Formula ClNa
Molecular Weight (g/mol) Mixture
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

CAS Min % 97.0, 0.98
CAS Max % 100.0, 1.02
Concentration 1% (w/w)
Packaging White Poly Wide Mouth Bottle
Solvent or Matrix Sodium Chloride
Color Gray
pH 7
Quantity 100 g
Physical Form Powder
Grade Indicator
Chemical Name or Material Mordant Black 11 Triturate, Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.