Learn More
Medchemexpress LLC Mordant orange 1 | 2243-76-7 | MFCD00007313 | 98.0% | 287.23 g/mol | C13H9N3O5 | 100 MG
SDP
Catalog No. 5000227574
Encompass_Preferred
Alizarine Yellow R (Mordant Orange 1) is an azo dye and salicylic acid derivative supplied as an orange to reddish powder. It is commonly used as a pH indicator and biological stain in chemical and histological applications, and is also employed in various dyeing processes.
  • Azo dye suitable as a pH indicator and histological stain.
  • Chemical formula C13H9N3O5; molecular weight 287.23 g/mol.
  • Supplied as a powder with a listed purity of 98.0%.
  • Available in small laboratory pack sizes, including 100 mg.
  • Used in laboratory staining, pH testing, and dyeing applications.
Catalog No. 50-002-27574 Supplier Medchemexpress LLC Supplier No. HY118320100MG
Add to Cart
Add to Cart
Alizarine Yellow R (Mordant Orange 1) is an azo dye and salicylic acid derivative supplied as an orange to reddish powder. It is commonly used as a pH indicator and biological stain in chemical and histological applications, and is also employed in various dyeing processes.
  • Azo dye suitable as a pH indicator and histological stain.
  • Chemical formula C13H9N3O5; molecular weight 287.23 g/mol.
  • Supplied as a powder with a listed purity of 98.0%.
  • Available in small laboratory pack sizes, including 100 mg.
  • Used in laboratory staining, pH testing, and dyeing applications.