Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mordant Yellow 10, pure, Thermo Scientific™

Catalog No. AC187180250
Change view
Click to view available options
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC187180250 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC187180250 Supplier Thermo Scientific Chemicals Supplier No. 187180250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6054-99-5
Dye Content 80% min.
Molecular Formula C13H8N2Na2O6S
Synonym C.I. 14010, 5-(4-Sulfophenylazo)salicylic acid, disodium salt
InChI Key CWQLKNYZZSWCNX-CBNWXSEKSA-L
SMILES C1=CC(=CC=C1NN=C2C=CC(=O)C(=C2)C(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
Infrared Spectrum Authentic
IUPAC Name disodium;(3E)-6-oxo-3-[(4-sulfonatophenyl)hydrazinylidene]cyclohexa-1,4-diene-1-carboxylate
Molecular Weight (g/mol) 366.255
PubChem CID 9575878
Formula Weight 366.26
Grade Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Brown to Yellow
Quantity 25g
Chemical Name or Material Mordant Yellow 10
Show More Show Less
Hazard Statement GHS H Statement:
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 227-976-2
RTECSNumber GB4450000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.