Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Morellic acid | 5304-71-2 | 5 MG
SDP

Catalog No. 5000418306
Encompass_Preferred

Morellic acid, isolated from Garcinia Morella, is an antiangiogenic agent. It inhibits the migration of HUVEC (human umbilical vein endothelial cells) at a low concentration of 0.5 μM in in vitro cell migration assays. It also demonstrates anticancer activity against human HT-29 cells with an ED50 of 0.36 μM.

  • Antiangiogenic activity
  • Inhibits HUVEC migration
  • Anticancer activity against HT-29 cells
  • Purity: 99.81%
  • Appearance: Orange to red solid
  • Soluble in DMSO (100 mg/mL)

Catalog No. 50-004-18306 Supplier Medchemexpress LLC Supplier No. HYN40945MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Morellic acid, isolated from Garcinia Morella, is an antiangiogenic agent. It inhibits the migration of HUVEC (human umbilical vein endothelial cells) at a low concentration of 0.5 μM in in vitro cell migration assays. It also demonstrates anticancer activity against human HT-29 cells with an ED50 of 0.36 μM.

  • Antiangiogenic activity
  • Inhibits HUVEC migration
  • Anticancer activity against HT-29 cells
  • Purity: 99.81%
  • Appearance: Orange to red solid
  • Soluble in DMSO (100 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.