Learn More
Aviva Systems Biology Corp MORF4L1 Antibody - middle region (ARP32832_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32832_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MORF4L1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human MORF4L1. Alternate names: Eaf3, MEAF3, MRG15, FWP006, S863-6, HsT17725, MORFRG15ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54781