Learn More
Aviva Systems Biology Corp Morf4l1 Antibody - middle region (ARP47607_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47607_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Morf4l1. It was validated on Western Blot. . Alternate names: KIAA4002, MGC102415, MGC103105, MGC118047, MORFRG15, MRG15, TEG-189, Tex189, mKIAA4002ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-315