Learn More
Aviva Systems Biology Corp MORF4L2 Antibody - N-terminal region (ARP33720_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33720_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MORF4L2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human MORF4L2. Alternate names: RP5-1055C14.2, KIAA0026, MORFL2, MRGXApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-014-55124