Learn More
ATCC Morganella morganii; genomic DNA
Catalog No. 502382253
Encompass
Genomic DNA isolated from Morganella morganii subsp. morganii strain AM-15 (ATCC 35200). This whole-genome sequenced product can be used in PCR and other molecular biology applications.
Catalog No. 50-238-2253 Supplier ATCC Supplier No. 35200D5
Genomic DNA isolated from Morganella morganii subsp. morganii strain AM-15 (ATCC 35200). This whole-genome sequenced product can be used in PCR and other molecular biology applications.