Learn More
ATCC Morganella morganii; subsp. morganii; Strain M11
Supplier:  ATCC 25830
Encompass
Morganella morganii subsp. morganii strain M11 was isolated from a patient with summer diarrhea. This whole-genome sequenced bacterium is a quality control strain for Sensititre products.
Catalog No. 50-238-4001