Learn More
Fine Science Tools Moria Iris Forceps, Straight/Smooth, 0.5 x 0.3 mm, 10 cm
SDP
Catalog No. NC0616811
Encompass
Style: MC32/B/ Tip Shape: Straight/ Tips: Smooth/ Tip Dimensions: 0.5 x 0.3mm/ Alloy / Material: Inox/ Length: 10cm
Catalog No. NC0616811 Supplier Fine Science Tools Supplier No. 1137322
Style: MC32/B/ Tip Shape: Straight/ Tips: Smooth/ Tip Dimensions: 0.5 x 0.3mm/ Alloy / Material: Inox/ Length: 10cm