Learn More
Fine Science Tools Moria Perforated Spoons, Tip: 15 mm, 14.5 cm
SDP
Catalog No. NC9190356
Encompass
Individual Item Name: Moria MC 17 BIS Perforated Spoon - 14.5cm/15mm Tip Diameter/ Tip Diameter: 15mm/ Spoon Depth: 3mm/ Alloy: Inox/ Length: 14.5cm
Catalog No. NC9190356 Supplier Fine Science Tools Supplier No. 1037018
Individual Item Name: Moria MC 17 BIS Perforated Spoon - 14.5cm/15mm Tip Diameter/ Tip Diameter: 15mm/ Spoon Depth: 3mm/ Alloy: Inox/ Length: 14.5cm